Search
Search

tpa-1| Gene Symbol: tpa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372683.1 | NP_001359808.1 | 11 - 12 | 2146 | 53 | |
| NM_001372684.2 | NP_001359809.1 | 11 - 12 | 2062 | 53 | ||
Set Membership: |
|