Search
Search

hecd-1| Gene Symbol: hecd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306759.2 | NP_001293688.1 | 9 - 10 | 2698 | 74 | |
| NM_001306760.2 | NP_001293689.1 | 9 - 10 | 2587 | 74 | ||
| NM_001380310.2 | NP_001368366.1 | 9 - 10 | 2594 | 74 | ||
| NM_001380312.2 | NP_001368365.1 | 9 - 10 | 2704 | 74 | ||
Set Membership: |
|