Search
Search

| Gene Symbol: dif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383120.1 | 6 - 7 | 2330 | 108 | ||
| Gene Symbol: F49E8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380331.2 | NP_001367809.1 | 6 - 7 | 2388 | 108 | |
Set Membership: |
|