Search
Search

ints-6| Gene Symbol: ints-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307283.3 | NP_001294212.1 | 14 - 15 | 2751 | 65 | |
| NM_001307284.2 | NP_001294213.1 | 14 - 15 | 2423 | 65 | ||
Set Membership: |
|