Search
Search

alp-1| Gene Symbol: alp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028199.5 | NP_001023370.1 | 11 - 12 | 1832 | 70 | |
| NM_001028200.6 | NP_001023371.1 | 13 - 14 | 3789 | 70 | ||
| NM_001047522.4 | NP_001040987.1 | 11 - 12 | 1575 | 70 | ||
| NM_069133.5 | NP_501534.2 | 10 - 11 | 1452 | 70 | ||
Set Membership: |
|