Search
Search

nhr-11| Gene Symbol: nhr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047598.3 | NP_001041063.1 | 7 - 8 | 888 | 128 | |
| NM_001047599.4 | NP_001041064.2 | 4 - 5 | 537 | 128 | ||
| NM_001268430.4 | NP_001255359.1 | 7 - 8 | 886 | 128 | ||
| NM_001268431.3 | NP_001255360.1 | 3 - 4 | 342 | 128 | ||
| NR_003464.1 | 3 - 4 | 845 | 128 | |||
| GenBank mRNA | AY305838.1 | - | 7 - 8 | 910 | 128 | |
Set Membership: |
|