Search
Search

fbl-1| Gene Symbol: fbl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028065.3 | NP_001023236.1 | 3 - 4 | 476 | 67 | |
| NM_001028066.6 | NP_001023237.1 | 3 - 4 | 449 | 67 | ||
| NM_001306946.3 | NP_001293875.1 | 3 - 4 | 432 | 67 | ||
| NM_001306947.3 | NP_001293876.1 | 3 - 4 | 432 | 67 | ||
| NM_001306948.3 | NP_001293877.1 | 3 - 4 | 432 | 67 | ||
| NM_001306949.2 | NP_001293878.1 | 3 - 4 | 476 | 67 | ||
| NM_001306950.3 | NP_001293879.1 | 3 - 4 | 432 | 67 | ||
| NM_069293.6 | NP_501694.2 | 3 - 4 | 478 | 67 | ||
| GenBank mRNA | AB212860.1 | - | 3 - 4 | 478 | 67 | |
| AF051401.1 | - | 3 - 4 | 432 | 67 | ||
| AF051402.1 | - | 3 - 4 | 432 | 67 | ||
| AY851363.1 | - | 3 - 4 | 432 | 67 | ||
Set Membership: |
|