Search
Search

catp-6| Gene Symbol: catp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268501.2 | NP_001255430.2 | 19 - 20 | 3492 | 137 | |
| NM_001268502.3 | NP_001255431.1 | 18 - 19 | 3312 | 137 | ||
| NM_001268503.3 | NP_001255432.1 | 11 - 12 | 2370 | 137 | ||
Set Membership: |
|