Search
Search

abt-1| Gene Symbol: abt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306925.2 | NP_001293854.1 | 27 - 28 | 4578 | 66 | |
| NM_001306926.3 | NP_001293855.1 | 24 - 25 | 4341 | 66 | ||
Set Membership: |
|