Search
Search

ugt-43| Gene Symbol: ugt-43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268558.2 | NP_001255487.1 | 4 - 5 | 963 | 76 | |
| NM_001268559.3 | NP_001255488.1 | 4 - 5 | 893 | 76 | ||
Set Membership: |
|