Search
Search

ubxn-2| Gene Symbol: ubxn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028419.5 | NP_001023590.1 | 5 - 6 | 762 | 83 | |
| NM_001380105.1 | NP_001368066.1 | 3 - 4 | 438 | 83 | ||
Set Membership: |
|