Search
Search

dyf-3| Gene Symbol: dyf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026125.3 | NP_001021296.1 | 3 - 4 | 553 | 69 | |
| NM_001380144.1 | NP_001367378.1 | 4 - 5 | 586 | 69 | ||
| NM_001380145.1 | NP_001367377.1 | 4 - 5 | 694 | 69 | ||
Set Membership: |
|