Search
Search

otpl-1| Gene Symbol: otpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307511.1 | NP_001294440.1 | 6 - 7 | 690 | 65 | |
| NM_001380150.1 | NP_001368270.1 | 7 - 8 | 750 | 65 | ||
| NM_067973.4 | NP_500374.2 | 2 - 3 | 302 | 65 | ||
Set Membership: |
|