Search
Search

psr-1| Gene Symbol: psr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047475.5 | NP_001040940.1 | 5 - 6 | 1036 | 88 | |
| NM_001047476.5 | NP_001040941.1 | 2 - 3 | 609 | 88 | ||
| NM_001307453.3 | NP_001294382.1 | 5 - 6 | 1005 | 88 | ||
Set Membership: |
|