Search
Search

smg-3| Gene Symbol: smg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380273.1 | NP_001368064.1 | 7 - 8 | 2198 | 117 | |
| NM_068573.9 | NP_500974.2 | 7 - 8 | 2202 | 117 | ||
| GenBank mRNA | EF472000.1 | - | 7 - 8 | 2216 | 117 | |
Set Membership: |
|