Search
Search

nhx-9| Gene Symbol: nhx-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028456.4 | NP_001023627.1 | 8 - 9 | 2199 | 83 | |
| NM_001028457.5 | NP_001023628.1 | 8 - 9 | 1877 | 83 | ||
| NM_001383212.1 | 8 - 9 | 2009 | 83 | |||
| GenBank mRNA | AF497834.1 | - | 8 - 9 | 1899 | 83 | |
| AF497835.1 | - | 8 - 9 | 2151 | 83 | ||
Set Membership: |
|