Search
Search

vha-7| Gene Symbol: vha-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383222.1 | 7 - 8 | 1294 | 64 | ||
| NM_070018.6 | NP_502419.2 | 2 - 3 | 562 | 64 | ||
| GenBank mRNA | AB055112.1 | - | 4 - 5 | 959 | 64 | |
Set Membership: |
|