Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268734.3 | NP_001255663.1 | 12 - 13 | 2238 | 136 | |
NM_001268735.4 | NP_001255664.1 | 11 - 12 | 2217 | 136 | ||
NM_001268736.3 | NP_001255665.1 | 10 - 11 | 2181 | 136 | ||
NM_001268737.4 | NP_001255666.1 | 12 - 13 | 2272 | 136 | ||
NM_001268738.3 | NP_001255667.1 | 11 - 12 | 2241 | 136 | ||
NM_001268739.3 | NP_001255668.1 | 3 - 4 | 873 | 136 | ||
NM_001268740.3 | NP_001255669.1 | 3 - 4 | 873 | 136 | ||
NM_001306972.3 | NP_001293901.1 | 13 - 14 | 2274 | 136 | ||
NM_001306973.3 | NP_001293902.1 | 13 - 14 | 2274 | 136 | ||
NM_001306974.3 | NP_001293903.1 | 12 - 13 | 2253 | 136 | ||
NM_001306975.3 | NP_001293904.1 | 12 - 13 | 2253 | 136 | ||
NM_001306976.3 | NP_001293905.1 | 11 - 12 | 2217 | 136 | ||
NM_001306977.3 | NP_001293906.1 | 11 - 12 | 2217 | 136 | ||
NM_001306978.3 | NP_001293907.1 | 12 - 13 | 2238 | 136 | ||
NM_001306979.3 | NP_001293908.1 | 11 - 12 | 2217 | 136 | ||
NM_001306980.3 | NP_001293909.1 | 10 - 11 | 2181 | 136 | ||
NM_001306981.3 | NP_001293910.1 | 3 - 4 | 873 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |