Search
Search

unc-30| Gene Symbol: unc-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026105.4 | NP_001021276.1 | 5 - 6 | 867 | 86 | |
| NM_001026106.2 | NP_001021277.1 | 5 - 6 | 754 | 86 | ||
| NM_001268771.3 | NP_001255700.1 | 2 - 3 | 216 | 86 | ||
| NM_001268772.4 | NP_001255701.1 | 5 - 6 | 599 | 86 | ||
Set Membership: |
|