Search
Search

rec-8| Gene Symbol: rec-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268798.2 | NP_001255727.1 | 5 - 6 | 2014 | 86 | |
| NM_001268799.3 | NP_001255728.1 | 1 - 2 | 1432 | 86 | ||
Set Membership: |
|