Search
Search

aptf-2| Gene Symbol: aptf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268811.3 | NP_001255740.1 | 3 - 4 | 946 | 80 | |
| NM_001268812.3 | NP_001255741.1 | 2 - 3 | 837 | 80 | ||
Set Membership: |
|