Search
Search

rsd-2| Gene Symbol: rsd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047487.3 | NP_001040952.2 | 19 - 20 | 3369 | 100 | |
| NM_001047488.4 | NP_001040953.1 | 16 - 17 | 2799 | 100 | ||
| NM_001268820.3 | NP_001255749.1 | 12 - 13 | 2025 | 100 | ||
| NM_001268821.3 | NP_001255750.1 | 4 - 5 | 663 | 100 | ||
Set Membership: |
|