Search
Search

tag-273| Gene Symbol: tag-273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028345.5 | NP_001023516.1 | 1 - 2 | 312 | 67 | |
| NM_001028346.4 | NP_001023517.2 | 7 - 8 | 1002 | 67 | ||
| NM_001268884.3 | NP_001255813.1 | 7 - 8 | 975 | 67 | ||
| NM_001307071.3 | NP_001294000.1 | 8 - 9 | 999 | 67 | ||
| NM_001307072.3 | NP_001294001.1 | 7 - 8 | 975 | 67 | ||
Set Membership: |
|