Search
Search

rho-1| Gene Symbol: rho-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070558.8 | NP_502959.1 | 2 - 3 | 221 | 86 | |
| NR_131670.1 | 1 - 2 | 138 | 86 | |||
| GenBank mRNA | L36965.1 | - | 2 - 3 | 223 | 86 | |
Set Membership: |
|