Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rom-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047548.5 | NP_001041013.2 | 12 - 13 | 3180 | 122 | |
NM_001083250.6 | NP_001076719.2 | 12 - 13 | 3186 | 122 | ||
NM_001380564.1 | NP_001368544.1 | 5 - 6 | 2235 | 122 | ||
NM_001380565.1 | NP_001368545.1 | 3 - 4 | 1389 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |