Search
Search

anat-1| Gene Symbol: anat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083193.5 | NP_001076662.1 | 5 - 6 | 648 | 64 | |
| NM_001083194.4 | NP_001076663.1 | 3 - 4 | 485 | 64 | ||
Set Membership: |
|