Search
Search

lec-12| Gene Symbol: lec-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028587.4 | NP_001023758.2 | 3 - 4 | 266 | 124 | |
| NM_001028588.5 | NP_001023759.2 | 3 - 4 | 266 | 124 | ||
| NM_001313136.3 | NP_001300065.1 | 2 - 3 | 138 | 124 | ||
| NM_001313137.3 | NP_001300066.1 | 2 - 3 | 138 | 124 | ||
Set Membership: |
|