Search
Search

unc-34| Gene Symbol: unc-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029083.5 | NP_001024254.1 | 1 - 2 | 346 | 115 | |
| NM_001313344.4 | NP_001300273.1 | 2 | 388 | 115 | ||
| NM_001313345.3 | NP_001300274.1 | 1 - 2 | 358 | 115 | ||
| NM_070959.8 | NP_503360.4 | 1 - 2 | 388 | 115 | ||
| GenBank mRNA | AF535160.1 | - | 2 - 3 | 583 | 115 | |
Set Membership: |
|