Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313356.3 | NP_001300285.1 | 3 - 4 | 1150 | 132 | |
NM_001313357.3 | NP_001300286.1 | 2 - 3 | 879 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |