Search
Search

ketn-1| Gene Symbol: ketn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307737.5 | NP_001294666.1 | 20 - 21 | 5681 | 70 | |
| NM_001307738.4 | NP_001294667.1 | 20 - 21 | 5681 | 70 | ||
| NM_001307739.4 | NP_001294668.1 | 20 - 21 | 5681 | 70 | ||
| NM_001307740.4 | NP_001294669.1 | 20 - 21 | 5681 | 70 | ||
| NM_001316807.3 | NP_001303736.1 | 7 - 8 | 1205 | 70 | ||
| NM_001316808.3 | NP_001303737.1 | 4 - 5 | 794 | 70 | ||
| NM_001377590.2 | NP_001364781.1 | 12 - 13 | 3764 | 70 | ||
| NM_001377591.2 | NP_001364828.1 | 12 - 13 | 3764 | 70 | ||
| NM_001377592.1 | NP_001364782.1 | 12 - 13 | 3764 | 70 | ||
| NM_001377593.1 | NP_001364783.1 | 12 - 13 | 3764 | 70 | ||
| NM_001377594.2 | NP_001364827.1 | 12 - 13 | 3764 | 70 | ||
| GenBank mRNA | AY819766.1 | - | 15 - 16 | 4551 | 70 | |
Set Membership: |
|