Search
Search

klc-2| Gene Symbol: klc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028492.4 | NP_001023663.1 | 4 - 5 | 1480 | 68 | |
| NM_001028493.6 | NP_001023664.1 | 4 - 5 | 1489 | 68 | ||
| NM_001028494.5 | NP_001023665.1 | 3 - 4 | 1386 | 68 | ||
Set Membership: |
|