Search
Search

mcp-1| Gene Symbol: mcp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028468.4 | NP_001023639.1 | 3 - 4 | 294 | 104 | |
| NM_001380650.1 | NP_001367490.1 | 3 - 4 | 361 | 104 | ||
Set Membership: |
|