Search
Search

| Gene Symbol: T25F10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072309.3 | NP_504710.1 | 9 - 10 | 1112 | 137 | |
| Gene Symbol: bbs-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072310.6 | NP_504711.2 | 16 - 17 | 2966 | 137 | |
Set Membership: |
|