Search
Search

acr-16| Gene Symbol: acr-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072806.5 | NP_505207.1 | 4 - 5 | 652 | 104 | |
| GenBank mRNA | AY523511.1 | - | 4 - 5 | 652 | 104 | |
| X83887.1 | - | 639 | 104 | |||
Set Membership: |
|