Search
Search

smk-1| Gene Symbol: smk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028738.7 | NP_001023909.2 | 7 - 8 | 1872 | 142 | |
| NM_001028739.6 | NP_001023910.2 | 4 - 5 | 1260 | 142 | ||
| NR_138495.1 | 6 - 7 | 1916 | 142 | |||
Set Membership: |
|