Search
Search

hum-2| Gene Symbol: hum-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330875.4 | NP_001317742.1 | 16 - 17 | 5360 | 81 | |
| NM_001330876.2 | NP_001317741.1 | 16 - 17 | 5354 | 81 | ||
| NM_001330877.3 | NP_001317812.1 | 10 - 11 | 2371 | 81 | ||
| NM_001330878.3 | NP_001317813.1 | 10 - 11 | 2365 | 81 | ||
| NM_001330879.3 | NP_001317814.1 | 9 - 10 | 2254 | 81 | ||
| NM_001330880.3 | NP_001317815.1 | 9 - 10 | 2248 | 81 | ||
| NM_001330881.3 | NP_001317816.1 | 8 - 9 | 2206 | 81 | ||
| NM_001373073.2 | NP_001359965.1 | 17 - 18 | 5281 | 81 | ||
| NM_001380727.2 | NP_001367720.1 | 16 - 17 | 5351 | 81 | ||
| NM_171505.6 | NP_741590.1 | 6 - 7 | 1449 | 81 | ||
Set Membership: |
|