Search
Search

dop-2| Gene Symbol: dop-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028876.3 | NP_001024047.1 | 4 - 5 | 548 | 70 | |
| NM_001269197.4 | NP_001256126.1 | 4 - 5 | 554 | 70 | ||
| NM_001269198.4 | NP_001256127.1 | 4 - 5 | 549 | 70 | ||
| NM_001373076.2 | NP_001360549.1 | 4 | 530 | 70 | ||
| NM_001373077.2 | NP_001360548.1 | 4 | 530 | 70 | ||
| NM_001373078.2 | NP_001360550.1 | 4 - 5 | 546 | 70 | ||
| GenBank mRNA | AB125702.1 | - | 4 - 5 | 570 | 70 | |
Set Membership: |
|