Search
Search

lsy-12| Gene Symbol: lsy-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269216.3 | NP_001256145.1 | 1 - 2 | 200 | 88 | |
| NM_001269218.3 | NP_001256147.1 | 1 - 2 | 190 | 88 | ||
| NM_001276918.3 | NP_001263847.1 | 1 - 2 | 190 | 88 | ||
| NM_001276920.4 | NP_001263849.1 | 1 - 2 | 190 | 88 | ||
Set Membership: |
|