Search
Search

vps-54| Gene Symbol: vps-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269297.2 | NP_001256226.1 | 7 - 8 | 2686 | 83 | |
| NM_001269298.3 | NP_001256227.1 | 2 - 3 | 1746 | 83 | ||
Set Membership: |
|