Search
Search

ceh-22| Gene Symbol: ceh-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083273.4 | NP_001076742.1 | 6 - 7 | 1545 | 74 | |
| NM_001083274.3 | NP_001076743.1 | 3 - 4 | 436 | 74 | ||
| NM_001083275.4 | NP_001076744.1 | 5 - 6 | 640 | 74 | ||
| NM_001356738.3 | NP_001343841.1 | 7 - 8 | 928 | 74 | ||
| NM_001356739.4 | NP_001343842.1 | 5 | 650 | 74 | ||
Set Membership: |
|