Search
Search

lact-8| Gene Symbol: lact-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269357.2 | NP_001256286.1 | 1 - 2 | 360 | 105 | |
| NM_001269358.3 | NP_001256287.1 | 1 - 2 | 86 | 105 | ||
Set Membership: |
|