Search
Search

snf-11| Gene Symbol: snf-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073472.7 | NP_505873.2 | 3 - 4 | 625 | 94 | |
| GenBank mRNA | AY529124.1 | - | 3 - 4 | 637 | 94 | |
| DQ482732.1 | - | 3 - 4 | 638 | 94 | ||
Set Membership: |
|