Search
Search

inx-11| Gene Symbol: inx-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269497.3 | NP_001256426.1 | 7 - 8 | 1179 | 94 | |
| NM_001373129.3 | NP_001359736.1 | 9 | 1206 | 94 | ||
| GenBank mRNA | KF137645.1 | - | 8 - 9 | 1174 | 94 | |
Set Membership: |
|