Search
Search

ril-2| Gene Symbol: ril-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028487.7 | NP_001023658.1 | 5 - 6 | 1714 | 125 | |
| NM_001028488.6 | NP_001023659.1 | 5 - 6 | 1716 | 125 | ||
| NM_001269507.4 | NP_001256436.1 | 5 - 6 | 1720 | 125 | ||
Set Membership: |
|