Search
Search

gpa-13| Gene Symbol: gpa-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269521.2 | NP_001256450.1 | 2 - 3 | 127 | 102 | |
| NM_001269522.3 | NP_001256451.1 | 3 - 4 | 335 | 102 | ||
| GenBank mRNA | AY008135.1 | - | 2 - 3 | 127 | 102 | |
Set Membership: |
|