Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gut-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269582.2 | NP_001256511.1 | 3 - 4 | 294 | 101 | |
NM_001269583.3 | NP_001256512.1 | 1 - 2 | 97 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |