Search
Search

avr-15| Gene Symbol: avr-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028905.8 | NP_001024076.1 | 5 - 6 | 1484 | 99 | |
| NM_001028906.5 | NP_001024077.1 | 3 - 4 | 928 | 99 | ||
Set Membership: |
|