Search
Search

sel-10| Gene Symbol: sel-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028804.4 | NP_001023975.1 | 7 - 8 | 729 | 100 | |
| NM_074020.7 | NP_506421.1 | 7 - 8 | 730 | 100 | ||
| NR_003169.1 | 6 - 7 | 765 | 100 | |||
| GenBank mRNA | AF020788.1 | - | 6 - 7 | 653 | 100 | |
Set Membership: |
|