Search
Search

nep-21| Gene Symbol: nep-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276955.4 | NP_001263884.1 | 10 - 11 | 1984 | 91 | |
| NM_001356755.4 | NP_001343820.1 | 11 - 12 | 2033 | 91 | ||
| NM_001373156.2 | NP_001360525.1 | 11 - 12 | 2061 | 91 | ||
| NM_074119.7 | NP_506520.1 | 10 - 11 | 1953 | 91 | ||
Set Membership: |
|