Search
Search

| Gene Symbol: srm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074154.5 | NP_506555.2 | 4 - 5 | 856 | 105 | |
| Gene Symbol: srm-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028771.4 | NP_001023942.1 | 9 - 10 | 2203 | 105 | |
Set Membership: |
|